Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl α-D-galactopyranoside | 3396-99-4 | 99.6% | 194.18 | C7H14O6 | 10 G
SDP

Catalog No. 5000208546
Encompass_Preferred

Methyl α-D-galactopyranoside is a methyl glycoside of D-galactose used as a biochemical reagent in glycobiology research and assays. It is typically supplied as a solid suitable for laboratory use.

  • Purity 99.58%.
  • Molecular weight 194.18 g/mol.
  • Chemical formula C7H14O6.
  • CAS number 3396-99-4.
  • Solid appearance, suitable for biochemical assays and glycobiology applications.

Catalog No. 50-002-08546 Supplier Medchemexpress LLC Supplier No. HYW08823810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl α-D-galactopyranoside is a methyl glycoside of D-galactose used as a biochemical reagent in glycobiology research and assays. It is typically supplied as a solid suitable for laboratory use.

  • Purity 99.58%.
  • Molecular weight 194.18 g/mol.
  • Chemical formula C7H14O6.
  • CAS number 3396-99-4.
  • Solid appearance, suitable for biochemical assays and glycobiology applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.