Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl α-D-galactopyranoside | 3396-99-4 | MFCD00006606 | 194.18 g/mol | C7H14O6 | 100 G
SDP

Catalog No. 5000208452
Encompass_Preferred

Methyl α-D-galactopyranoside is a methyl glycoside of D-galactopyranose used as a biochemical reagent in glycobiology and carbohydrate research. It is a solid with molecular formula C7H14O6 and molecular weight 194.18 g/mol. Refer to the Safety Data Sheet for handling, storage, and safety information.

  • Methyl glycoside of D-galactopyranose
  • Used in glycobiology and carbohydrate research
  • Solid; melting point 116-117 °C
  • Molecular formula C7H14O6
  • Molecular weight 194.18 g/mol
  • Recommended storage: powder -20 °C (3 years); 4 °C (2 years)

Catalog No. 50-002-08452 Supplier Medchemexpress LLC Supplier No. HYW088238100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl α-D-galactopyranoside is a methyl glycoside of D-galactopyranose used as a biochemical reagent in glycobiology and carbohydrate research. It is a solid with molecular formula C7H14O6 and molecular weight 194.18 g/mol. Refer to the Safety Data Sheet for handling, storage, and safety information.

  • Methyl glycoside of D-galactopyranose
  • Used in glycobiology and carbohydrate research
  • Solid; melting point 116-117 °C
  • Molecular formula C7H14O6
  • Molecular weight 194.18 g/mol
  • Recommended storage: powder -20 °C (3 years); 4 °C (2 years)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.