Learn More
Medchemexpress LLC Methyl α-D-galactopyranoside | 3396-99-4 | MFCD64085 | >98.0% | 194.18 g·mol⁻1 | C7H14O6 | 50 G
SDP
Catalog No. 5000208455
Encompass_Preferred
Methyl α-D-galactopyranoside is the methyl glycoside of D-galactopyranose used as a reagent in glycobiology and enzymology research. It functions as a substrate or inhibitor in studies of galactosidase enzymes and carbohydrate metabolism. Typical properties include molecular formula C7H14O6 and molecular weight 194.18 g·mol⁻1.
  • Small-molecule carbohydrate reagent for research use.
  • Used as a substrate or inhibitor for galactosidase assays.
  • Molecular formula C7H14O6; molecular weight 194.18 g·mol⁻1.
  • Reported purity >98.0% (vendor specifications).
  • Available as a 50 g powder package.
Catalog No. 50-002-08455 Supplier Medchemexpress LLC Supplier No. HYW08823850G
Methyl α-D-galactopyranoside is the methyl glycoside of D-galactopyranose used as a reagent in glycobiology and enzymology research. It functions as a substrate or inhibitor in studies of galactosidase enzymes and carbohydrate metabolism. Typical properties include molecular formula C7H14O6 and molecular weight 194.18 g·mol⁻1.
  • Small-molecule carbohydrate reagent for research use.
  • Used as a substrate or inhibitor for galactosidase assays.
  • Molecular formula C7H14O6; molecular weight 194.18 g·mol⁻1.
  • Reported purity >98.0% (vendor specifications).
  • Available as a 50 g powder package.