Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl α-D-glucopyranoside | 97-30-3 | MFCD00064086 | ≥98.0% | 194.18 g/mol | C7H14O6 | 100 G
SDP

Catalog No. 5000208702
Encompass_Preferred

Methyl α-D-glucopyranoside is a methylated glucose derivative used as a reagent in glycosylation studies and biochemical assays. It is supplied as a solid with accompanying documentation for purity and safety.

  • Chemical name: methyl α-D-glucopyranoside.
  • Cas number: 97-30-3.
  • Molecular formula: C7H14O6.
  • Molecular weight: 194.18 g/mol.
  • Purity: ≥98.0%.
  • Physical form: solid (powder).
  • Available package sizes: 25 g, 50 g, 100 g, bulk quantities available on request.
  • Documentation: datasheet, certificate of analysis, and safety data sheet available.

Catalog No. 50-002-08702 Supplier Medchemexpress LLC Supplier No. HYW040294100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl α-D-glucopyranoside is a methylated glucose derivative used as a reagent in glycosylation studies and biochemical assays. It is supplied as a solid with accompanying documentation for purity and safety.

  • Chemical name: methyl α-D-glucopyranoside.
  • Cas number: 97-30-3.
  • Molecular formula: C7H14O6.
  • Molecular weight: 194.18 g/mol.
  • Purity: ≥98.0%.
  • Physical form: solid (powder).
  • Available package sizes: 25 g, 50 g, 100 g, bulk quantities available on request.
  • Documentation: datasheet, certificate of analysis, and safety data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.