Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl α-D-glucopyranoside | 97-30-3 | MFCD00064086 | ≥98.0% | 194.18 g/mol | C7H14O6 | 50 G
SDP

Catalog No. 5000209564
Encompass_Preferred

Methyl α-D-glucopyranoside is a purified methyl glycoside of D-glucose used as a biochemical reagent for glycosylation studies, analytical assays, and general life-science research. It is supplied as a solid with high purity suitable for laboratory use.

  • Methyl α-D-glucopyranoside, CAS number 97-30-3.
  • Purity ≥98.0%.
  • Molecular formula C7H14O6.
  • Molecular weight 194.18 g/mol.
  • Available as a 50 G pack.

Catalog No. 50-002-09564 Supplier Medchemexpress LLC Supplier No. HYW04029450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl α-D-glucopyranoside is a purified methyl glycoside of D-glucose used as a biochemical reagent for glycosylation studies, analytical assays, and general life-science research. It is supplied as a solid with high purity suitable for laboratory use.

  • Methyl α-D-glucopyranoside, CAS number 97-30-3.
  • Purity ≥98.0%.
  • Molecular formula C7H14O6.
  • Molecular weight 194.18 g/mol.
  • Available as a 50 G pack.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.