Learn More
Alkali Scientific Methyl β D glucuronide Sodium Salt, 1 Gram
SDP
Catalog No. 5000660583
Encompass_Preferred
Methyl β-D-Glucuronide Sodium Salt (1g) is a glucuronide compound used in biochemical research and laboratory applications. This reagent is commonly used in assays to measure enzymatic activity, particularly in the context of glucuronidation. The sodium salt form enhances solubility and stability in aqueous solutions, making it suitable for use in enzymatic assays, drug metabolism studies, and various laboratory investigations. The compound is frequently employed in studies related to detoxification, metabolism, and cellular enzyme activity, offering reliable and reproducible results in experiments requiring high purity and sensitivity.
Catalog No. 50-006-60583 Supplier Alkali Scientific Supplier No. YLB001CP
Methyl β-D-Glucuronide Sodium Salt (1g) is a glucuronide compound used in biochemical research and laboratory applications. This reagent is commonly used in assays to measure enzymatic activity, particularly in the context of glucuronidation. The sodium salt form enhances solubility and stability in aqueous solutions, making it suitable for use in enzymatic assays, drug metabolism studies, and various laboratory investigations. The compound is frequently employed in studies related to detoxification, metabolism, and cellular enzyme activity, offering reliable and reproducible results in experiments requiring high purity and sensitivity.