Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+)-Methyl D-Lactate

Catalog No. 2777625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-776-25G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 27-776-25G Supplier Sigma Aldrich Supplier No. 2777625G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(+)-Methyl D-lactate can be used as a starting material in the asymmetric preparation of: Neomethymycin, a macrolide which contains 12-membered macrolactone that is used as a potent biological agent. PM-toxin A.It can also be used as a chiral pool along with D-(-)-tartaric acid in the stereoselective total synthesis of separacenes A and B via Trost-Rychnovsky alkyne rearrangement, Horner-Wadsworth-Emmons olefination as well as Corey-Bakshi-Shibata reaction.

Specifications

CAS 123334-22-5
Melting Point 220°C
Density 2.88 g/mL at 25°C
Molecular Formula C2H4Cl3KPt -+ xH2O
Refractive Index n20/D 1.413
Linear Formula K[(H2C=CH2)PtCl3] -+ xH2O
MDL Number MFCD00150524
Quantity 500 mg
Synonym D-Lactic acid methyl ester; Methyl (R)-(+)-lactate
Molecular Weight (g/mol) 368.59 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.