Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl D-(-)-Mandelate 98.0+%, TCI America™
SDP

Catalog No. M13491G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13491G 1 g
M13495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13491G Supplier TCI America Supplier No. M13491G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20698-91-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00064247
InChI Key ITATYELQCJRCCK-MRVPVSSYSA-N
Synonym methyl r---mandelate, r---methyl mandelate, r-methyl 2-hydroxy-2-phenylacetate, d---mandelic acid methyl ester, methyl 2r-2-hydroxy-2-phenylacetate, r-methyl mandelate, methyl d---mandelate, r-mandelic acid methyl ester, r---mandelic acid methyl ester, methyl hydroxy phenyl acetate #
PubChem CID 2724623
IUPAC Name methyl (2R)-2-hydroxy-2-phenylacetate
SMILES COC(=O)C(C1=CC=CC=C1)O

Specifications

Color White
Quantity 1 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl D-(-)-Mandelate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.