Learn More
Apexbio Technology LLC Methyl α-D-mannopyranoside 617-04-9 25g
SDP
Catalog No. 5000094511
Encompass_Preferred
Methyl alpha-D-mannopyranoside (CAS 617-04-9) is a small-molecule ligand targeting mannose receptors expressed on macrophages It is designed to bind selectively to these receptors thereby facilitating internalization and intracellular delivery of conjugated therapeutic agents Methyl alpha-D-mannopyranoside exerts its biological activity primarily through specific binding to macrophage mannose receptors enabling targeted transport into macrophages Studies evaluating receptor binding report IC50 values generally ranging from micromolar to millimolar levels depending on experimental conditions and assay methods Based on these pharmacological properties methyl alpha-D-mannopyranoside holds research potential in targeted drug delivery for anti-tubercular therapies
Catalog No. 50-000-94511 Supplier Apexbio Technology LLC Supplier No. A998825000
Methyl alpha-D-mannopyranoside (CAS 617-04-9) is a small-molecule ligand targeting mannose receptors expressed on macrophages It is designed to bind selectively to these receptors thereby facilitating internalization and intracellular delivery of conjugated therapeutic agents Methyl alpha-D-mannopyranoside exerts its biological activity primarily through specific binding to macrophage mannose receptors enabling targeted transport into macrophages Studies evaluating receptor binding report IC50 values generally ranging from micromolar to millimolar levels depending on experimental conditions and assay methods Based on these pharmacological properties methyl alpha-D-mannopyranoside holds research potential in targeted drug delivery for anti-tubercular therapies