Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl α-D-mannopyranoside | 617-04-9 | MFCD00063262 | 98.0% | 194.18 g/mol | C7H14O6 | 1 ML
SDP

Catalog No. 5000301215
Encompass_Preferred

Methyl α-D-mannopyranoside is a methylated mannose derivative used as a research reagent in glycobiology and microbiology for lectin-binding studies and as a competitive inhibitor of mannose-binding proteins. It is supplied as a white to off-white solid with high purity and defined storage recommendations.

  • Cas number 617-04-9.
  • Molecular formula C7H14O6.
  • Molecular weight 194.18 g/mol.
  • Purity 98.0% (NMR).
  • Appearance white to off-white solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Typical applications include glycobiology, lectin inhibition, and microbiology research.
  • For research use only; not for human therapeutic use.

Catalog No. 50-003-01215 Supplier Medchemexpress LLC Supplier No. HYW03989710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl α-D-mannopyranoside is a methylated mannose derivative used as a research reagent in glycobiology and microbiology for lectin-binding studies and as a competitive inhibitor of mannose-binding proteins. It is supplied as a white to off-white solid with high purity and defined storage recommendations.

  • Cas number 617-04-9.
  • Molecular formula C7H14O6.
  • Molecular weight 194.18 g/mol.
  • Purity 98.0% (NMR).
  • Appearance white to off-white solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Typical applications include glycobiology, lectin inhibition, and microbiology research.
  • For research use only; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.