Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl α-D-mannopyranoside | 617-04-9 | MFCD00063262 | 98.0% | 194.18 g·mol⁻¹ | C7H14O6 | 10 G
SDP

Catalog No. 5000301955
Encompass_Preferred

Methyl α-D-mannopyranoside is a methylated mannose derivative used as a biochemical reagent and analytical standard in glycobiology, microbiology, and related research. Supplied as a powder, the compound has documented molecular properties and recommended cold storage conditions for solid and solution stability.

  • High purity: 98.0%
  • Molecular formula C7H14O6
  • Molecular weight 194.18 g·mol⁻¹
  • Powder form with recommended cold storage for long-term stability
  • Suitable as an analytical standard and for glycobiology assays
  • Reported application in macrophage-targeting inhalation research
  • Available in multiple solid pack sizes for research use

Catalog No. 50-003-01955 Supplier Medchemexpress LLC Supplier No. HYW03989710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl α-D-mannopyranoside is a methylated mannose derivative used as a biochemical reagent and analytical standard in glycobiology, microbiology, and related research. Supplied as a powder, the compound has documented molecular properties and recommended cold storage conditions for solid and solution stability.

  • High purity: 98.0%
  • Molecular formula C7H14O6
  • Molecular weight 194.18 g·mol⁻¹
  • Powder form with recommended cold storage for long-term stability
  • Suitable as an analytical standard and for glycobiology assays
  • Reported application in macrophage-targeting inhalation research
  • Available in multiple solid pack sizes for research use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.