Learn More
Medchemexpress LLC Methyl α-D-mannopyranoside | 617-04-9 | MFCD00063262 | 98.0% | 194.18 g/mol | C7H14O6 | 50 G
SDP
Catalog No. 5000299827
Encompass_Preferred
Methyl α-D-mannopyranoside is a methylated monosaccharide used as a biochemical reagent and analytical standard. It functions as a competitive inhibitor of mannose-binding interactions and is applied in glycobiology, protein purification, and microbiology research; it has also been investigated for macrophage-targeting in inhalation therapy studies. Supplied as a solid with documented purity and storage recommendations for research use only.
  • Acts as a competitive inhibitor of mannose-binding proteins.
  • Used as an analytical standard in glycobiology and biochemical assays.
  • Suitable for studies of mannose-dependent microbial adherence and lectin interactions.
  • High purity: 98.0% (NMR).
  • Molecular weight 194.18 g/mol; chemical formula C7H14O6.
  • White to off-white solid with defined storage recommendations for long-term stability.
  • Intended for research and analytical applications; not for human therapeutic use.
Catalog No. 50-002-99827 Supplier Medchemexpress LLC Supplier No. HYW03989750G
Methyl α-D-mannopyranoside is a methylated monosaccharide used as a biochemical reagent and analytical standard. It functions as a competitive inhibitor of mannose-binding interactions and is applied in glycobiology, protein purification, and microbiology research; it has also been investigated for macrophage-targeting in inhalation therapy studies. Supplied as a solid with documented purity and storage recommendations for research use only.
  • Acts as a competitive inhibitor of mannose-binding proteins.
  • Used as an analytical standard in glycobiology and biochemical assays.
  • Suitable for studies of mannose-dependent microbial adherence and lectin interactions.
  • High purity: 98.0% (NMR).
  • Molecular weight 194.18 g/mol; chemical formula C7H14O6.
  • White to off-white solid with defined storage recommendations for long-term stability.
  • Intended for research and analytical applications; not for human therapeutic use.