Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl deacetylasperulosidate | 52613-28-2 | 404.37 | 25 MG
SDP

Catalog No. 5000389860
Encompass_Preferred

Methyl deacetylasperulosidate is an iridoid that demonstrates purgative effects in mice and contributes to lowering blood glucose levels in normal mice.

  • Iridoid compound
  • Shows purgative effects in mice
  • Lowers blood glucose levels in normal mice
  • Molecular formula: C₁₇H₂₄O₁₁
  • Appearance: Solid, white to off-white
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Isolated from Plants (Rubiaceae, Hedyotis diffusa Willd.)
  • For research use only

Catalog No. 50-003-89860 Supplier Medchemexpress LLC Supplier No. 25MGHYN1503
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl deacetylasperulosidate is an iridoid that demonstrates purgative effects in mice and contributes to lowering blood glucose levels in normal mice.

  • Iridoid compound
  • Shows purgative effects in mice
  • Lowers blood glucose levels in normal mice
  • Molecular formula: C₁₇H₂₄O₁₁
  • Appearance: Solid, white to off-white
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Isolated from Plants (Rubiaceae, Hedyotis diffusa Willd.)
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.