Learn More
Medchemexpress LLC Methyl decanoate | 110-42-9 | MFCD00009580 | 99.8% | 186.30 | C11H22O2 | 1 ML
SDP
Catalog No. 5000302136
Encompass_Preferred
Methyl decanoate is the methyl ester of decanoic (capric) acid supplied as a ready-to-use 10 mM solution in DMSO for research and analytical applications. The reagent is high purity and suitable for use as a biochemical assay reagent, reference standard, and in flavor and fragrance studies.
  • Ready-to-use 10 mM solution in DMSO.
  • High purity (99.83%) suitable for analytical use.
  • Useful as a biochemical assay reagent and reference standard.
  • Applicable to flavor and fragrance research and method development.
  • Available in solution and bulk liquid pack sizes for flexibility.
Catalog No. 50-003-02136 Supplier Medchemexpress LLC Supplier No. HYW01617210MM/1ML
Methyl decanoate is the methyl ester of decanoic (capric) acid supplied as a ready-to-use 10 mM solution in DMSO for research and analytical applications. The reagent is high purity and suitable for use as a biochemical assay reagent, reference standard, and in flavor and fragrance studies.
  • Ready-to-use 10 mM solution in DMSO.
  • High purity (99.83%) suitable for analytical use.
  • Useful as a biochemical assay reagent and reference standard.
  • Applicable to flavor and fragrance research and method development.
  • Available in solution and bulk liquid pack sizes for flexibility.