Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl decanoate | 110-42-9 | MFCD00009580 | 99.8% | 186.30 g·mol⁻¹ | C11H22O2 | 500 G
SDP

Catalog No. 5000302137
Encompass_Preferred

Methyl decanoate (CAS 110-42-9) is the methyl ester of decanoic (capric) acid, supplied as a colorless to light-yellow liquid with a fruity odor for use as a biochemical reagent and flavoring component in research. It has formula C11H22O2 and molecular weight 186.30 g·mol⁻¹, and is provided at high purity with recommended storage for stability.

  • High purity: 99.83% suitable for research and analytical work.
  • Physical state: liquid, colorless to light yellow, fruity odor.
  • Molecular formula C11H22O2 and molecular weight 186.30 g·mol⁻¹.
  • Density 0.871 g/cm³ for formulation and handling calculations.
  • Recommended storage: follow temperature guidelines to maintain stability.
  • For research use only; not for human or clinical use.

Catalog No. 50-003-02137 Supplier Medchemexpress LLC Supplier No. HYW016172500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl decanoate (CAS 110-42-9) is the methyl ester of decanoic (capric) acid, supplied as a colorless to light-yellow liquid with a fruity odor for use as a biochemical reagent and flavoring component in research. It has formula C11H22O2 and molecular weight 186.30 g·mol⁻¹, and is provided at high purity with recommended storage for stability.

  • High purity: 99.83% suitable for research and analytical work.
  • Physical state: liquid, colorless to light yellow, fruity odor.
  • Molecular formula C11H22O2 and molecular weight 186.30 g·mol⁻¹.
  • Density 0.871 g/cm³ for formulation and handling calculations.
  • Recommended storage: follow temperature guidelines to maintain stability.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.