Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl Diethylphosphonoacetate

Catalog No. 15876350G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-876-350G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 15-876-350G Supplier Sigma Aldrich Supplier No. 15876350G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant or reagent for: Synthesis of branches allylic fluoridesRegioselective Diels-Alder reactions to yield antibiotic building blocksSubstituted thiophenes and furans synthesis for treatment of type 2 diabetesStereoselective Diels-Alder cycloaddition reactionsSynthesis of pyridone alkaloids with neuritogenic activityModification of botulinum neurotoxin serotype A protease inhibitorsSynthesis of immunosuppressive cyclopentanediol derivatives

Specifications

CAS 34404-36-9
Melting Point 110°C to 120°C
Molecular Formula C23H31N3O8
Refractive Index n20/D 1.433
Linear Formula C23H31N3O8
MDL Number MFCD00037915
Quantity 50 g
Synonym Methyl P,P-diethylphosphonoacetate; (Methoxycarbonylmethyl) diethoxyphosphine oxide; NSC 147757; [(Methoxycarbonyl)methyl]phosphonic acid diethyl ester
Molecular Weight (g/mol) 477.51 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.