Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl difluoroacetate, 98%

Catalog No. AAB2219706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2219706 5 g
AAB2219714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2219706 Supplier Thermo Scientific Chemicals Supplier No. B2219706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 433-53-4 | C3H4F2O2 | 110.06 g/mol

A methyl difluoroacetate (MFA)-based LiPF6 solution was applied as an electrolyte to improve the thermal stability of Li-ion batteries. Methyl difluoroacetate is used in the inhibition of corrosion of aluminum cathode current collector for lithium ion cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A methyl difluoroacetate (MFA)-based LiPF6 solution was applied as an electrolyte to improve the thermal stability of Li-ion batteries. Methyl difluoroacetate is used in the inhibition of corrosion of aluminum cathode current collector for lithium ion cells.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 433-53-4
Molecular Formula C3H4F2O2
Molecular Weight (g/mol) 110.06
MDL Number MFCD00039276
InChI Key CSSYKHYGURSRAZ-UHFFFAOYSA-N
Synonym methyl difluoroacetate, methyl2,2-difluoroacetate, difluoroacetic acid methyl ester, acetic acid, difluoro-, methyl ester, methyldifluoroacetate, methyl difluoroacetate #, acmc-209ju0, methyl 2,2-difluoroethanoate
PubChem CID 79012
IUPAC Name methyl 2,2-difluoroacetate
SMILES COC(=O)C(F)F

Specifications

Density 1.272
Boiling Point 86°C to 87°C
Flash Point 19°C (66°F)
Refractive Index 1.335
Quantity 5 g
UN Number UN2924
Beilstein 1747534
Solubility Information Slightly soluble in water.
Formula Weight 110.06
Percent Purity 99%
Chemical Name or Material Methyl difluoroacetate
Show More Show Less
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 207-089-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.