Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Difluoroacetate 97.0+%, TCI America™
SDP

Catalog No. M202125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M202125G 25 g
M20215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M202125G Supplier TCI America Supplier No. M202125G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 433-53-4
Molecular Formula C3H4F2O2
Molecular Weight (g/mol) 110.06
MDL Number MFCD00039276
InChI Key CSSYKHYGURSRAZ-UHFFFAOYSA-N
Synonym methyl difluoroacetate, methyl2,2-difluoroacetate, difluoroacetic acid methyl ester, acetic acid, difluoro-, methyl ester, methyldifluoroacetate, methyl difluoroacetate #, acmc-209ju0, methyl 2,2-difluoroethanoate
PubChem CID 79012
IUPAC Name methyl 2,2-difluoroacetate
SMILES COC(=O)C(F)F

Specifications

Color Yellow
Boiling Point 86°C
Quantity 25 g
UN Number 2924
Formula Weight 110.06
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Difluoroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.