Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Dihydrojasmonate (cis- and trans- mixture) 95.0+%, TCI America™
SDP

Catalog No. D143125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D143125ML 25 mL
D143110ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D143125ML Supplier TCI America Supplier No. D143125ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 24851-98-7
Molecular Formula C13H22O3
Molecular Weight (g/mol) 226.316
MDL Number MFCD00151188
InChI Key KVWWIYGFBYDJQC-UHFFFAOYSA-N
Synonym 2-Amylcyclopentan-1-one-3-acetic Acid Methyl Ester, Dihydrojasmonic Acid Methyl Ester, Methyl 3-Oxo-2-pentyl-1-cyclopentaneacetate
PubChem CID 102861
IUPAC Name methyl 2-(3-oxo-2-pentylcyclopentyl)acetate
SMILES CCCCCC1C(CCC1=O)CC(=O)OC

Specifications

Color Yellow
Boiling Point 110°C
Quantity 25 mL
Formula Weight 226.32
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Dihydrojasmonate (cis- and trans- mixture)
EINECSNumber (3)-3309
RTECSNumber GY2453800
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.