Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl diphenylacetate, 98%

Catalog No. AAB2469422
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2469422 100 g
AAB2469406 5 g
AAB2469414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2469422 Supplier Thermo Scientific Chemicals Supplier No. B2469422
Add to Cart

CAS: 3469-00-9 | C15H14O2 | 226.28 g/mol

Methyl diphenylacetate is used as a chemical, organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl diphenylacetate is used as a chemical, organic intermediate.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3469-00-9
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.28
MDL Number MFCD00025869
InChI Key AORIUCNKPVHMTN-UHFFFAOYSA-N
Synonym methyl diphenylacetate, diphenylacetic acid, methyl ester, acetic acid, diphenyl-, methyl ester, benzeneacetic acid, .alpha.-phenyl-, methyl ester, methyldiphenylacetate, cbmicro_020304, acmc-209i9m, c6h5 2chcooch3, diphenylacetic acid methyl ester
PubChem CID 77019
IUPAC Name methyl 2,2-diphenylacetate
SMILES COC(=O)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 59°C to 61°C
Quantity 100 g
Solubility Information Soluble in methanol.
Formula Weight 226.28
Percent Purity 98%
Chemical Name or Material Methyl diphenylacetate
EINECSNumber 222-431-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.