Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl DL-2-Aminobutyrate Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A08935G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08935G 5 g
A089325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A08935G Supplier TCI America Supplier No. A08935G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7682-18-0
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.606
MDL Number MFCD00058295
InChI Key AHAQQEGUPULIOZ-UHFFFAOYSA-N
Synonym DL-2-Aminobutyric Acid Methyl Ester Hydrochloride
PubChem CID 13283515
IUPAC Name methyl 2-aminobutanoate;hydrochloride
SMILES CCC(C(=O)OC)N.Cl

Specifications

Melting Point 150°C
Color White
Quantity 5 g
Formula Weight 153.61
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Methyl DL-2-Aminobutyrate Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.