Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl DL-Mandelate 98.0+%, TCI America™
SDP

Catalog No. M0040500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0040500G 500 g
M004025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0040500G Supplier TCI America Supplier No. M0040500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4358-87-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00004493
InChI Key ITATYELQCJRCCK-UHFFFAOYNA-N
Synonym methyl mandelate, methyl dl-mandelate, mandelic acid, methyl ester, methyl phenylglycolate, methyl hydroxy phenyl acetate, +/--methyl mandelate, mandelic acid methyl ester, dl-mandelic acid methyl ester, dl-mandelic acid, methyl ester, methyl 2-hydroxy-2-phenyl-acetate
PubChem CID 78066
IUPAC Name methyl 2-hydroxy-2-phenylacetate
SMILES COC(=O)C(O)C1=CC=CC=C1

Specifications

Melting Point 56°C
Color White
Boiling Point 250°C
Quantity 500 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl DL-Mandelate
EINECSNumber (3)-3842
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.