Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl DL-pyroglutamate | 54571-66-3 | 97.37% | 143.14 | 25 G
SDP

Catalog No. 5000403329
Encompass_Preferred

Methyl DL-pyroglutamate is a biochemical reagent intended for research use only. It is not sold to patients.

  • Appearance: Solid below 21°C, liquid above 23°C
  • Color: Colorless to light yellow
  • Shipping: Room temperature in continental US
  • Storage: Store at room temperature for 3 years; in solvent: -80°C for 2 years, -20°C for 1 year

Catalog No. 50-004-03329 Supplier Medchemexpress LLC Supplier No. 25GHY32290
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Methyl DL-pyroglutamate is a biochemical reagent intended for research use only. It is not sold to patients.

  • Appearance: Solid below 21°C, liquid above 23°C
  • Color: Colorless to light yellow
  • Shipping: Room temperature in continental US
  • Storage: Store at room temperature for 3 years; in solvent: -80°C for 2 years, -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.