Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl DL-pyroglutamate | 54571-66-3 | 97.4% | 143.14 | 50 G
SDP

Catalog No. 5000401995
Encompass_Preferred

Methyl DL-pyroglutamate is a biochemical reagent. It is intended for research use only and is not sold to patients.

  • Purity of 97.37%
  • Molecular weight of 143.14
  • CAS number: 54571-66-3
  • Chemical formula: C6H9NO3
  • Appearance: solid below 21°C, liquid above 23°C
  • Color: colorless to light yellow
  • Soluble in DMSO at 200 mg/mL

Catalog No. 50-004-01995 Supplier Medchemexpress LLC Supplier No. HY3229050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl DL-pyroglutamate is a biochemical reagent. It is intended for research use only and is not sold to patients.

  • Purity of 97.37%
  • Molecular weight of 143.14
  • CAS number: 54571-66-3
  • Chemical formula: C6H9NO3
  • Appearance: solid below 21°C, liquid above 23°C
  • Color: colorless to light yellow
  • Soluble in DMSO at 200 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.