Learn More
Medchemexpress LLC Methyl docosanoate | 929-77-1 | 354.62 | 100 MG
SDP
Catalog No. 5000451225
Encompass_Preferred
Methyl behenate (Standard) is the analytical standard of Methyl behenate (HY-W009082). This product is intended for research and analytical applications. Methyl behenate (Methyl docosanoate) is a fatty acid methyl ester. Methyl behenate can be isolated from the plant Aspidopterys obcordata Lemsl. Methyl behenate can be used as a phase change material.
  • Analytical standard for research and analytical applications
  • Can be isolated from the plant Aspidopterys obcordata Lemsl
  • Can be used as a phase change material
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
Catalog No. 50-004-51225 Supplier Medchemexpress LLC Supplier No. HYW009082100MG
Methyl behenate (Standard) is the analytical standard of Methyl behenate (HY-W009082). This product is intended for research and analytical applications. Methyl behenate (Methyl docosanoate) is a fatty acid methyl ester. Methyl behenate can be isolated from the plant Aspidopterys obcordata Lemsl. Methyl behenate can be used as a phase change material.
  • Analytical standard for research and analytical applications
  • Can be isolated from the plant Aspidopterys obcordata Lemsl
  • Can be used as a phase change material
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS