Learn More
Medchemexpress LLC Methyl (E)-cinnamate | 1754-62-7 | 97.6% | 162.19 g/mol | C10H10O2 | 500 G
SDP
Catalog No. 5000210466
Encompass_Preferred
Methyl (E)-cinnamate is a naturally occurring phytochemical and flavor compound supplied for research and analytical use. It is reported to have anti-inflammatory properties and is used in flavoring and commodity applications. Key identifiers include CAS number 1754-62-7, molecular formula C10H10O2, molecular weight 162.19 g/mol, and typical purity 97.6%.
  • Natural flavor compound with reported anti-inflammatory properties.
  • Available in 100 g, 500 g, and 1 kg pack sizes.
  • Typical purity 97.6%.
  • Molecular weight 162.19 g/mol, formula C10H10O2.
  • Intended for research and analytical use.
Catalog No. 50-002-10466 Supplier Medchemexpress LLC Supplier No. HYW067056500G
Methyl (E)-cinnamate is a naturally occurring phytochemical and flavor compound supplied for research and analytical use. It is reported to have anti-inflammatory properties and is used in flavoring and commodity applications. Key identifiers include CAS number 1754-62-7, molecular formula C10H10O2, molecular weight 162.19 g/mol, and typical purity 97.6%.
  • Natural flavor compound with reported anti-inflammatory properties.
  • Available in 100 g, 500 g, and 1 kg pack sizes.
  • Typical purity 97.6%.
  • Molecular weight 162.19 g/mol, formula C10H10O2.
  • Intended for research and analytical use.