Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl elaidate | 1937-62-8 | 1 ML
SDP

Catalog No. 5000418710
Encompass_Preferred

Methyl elaidate is a fatty acid ester derived from elaidic acid, a monounsaturated omega-9 fatty acid found in certain vegetable oils and animal fats. It is commonly used as a reference compound for the analysis of fatty acids and their derivatives by gas chromatography. This product is intended for research use only.

  • Derived from elaidic acid
  • Monounsaturated omega-9 fatty acid
  • Used as a reference compound for fatty acid analysis
  • Suitable for gas chromatography
  • Purity: 99.22%
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • For research use only
  • Store neat at 4°C under nitrogen
  • Store in solvent at -80°C for 6 months or -20°C for 1 month under nitrogen

Catalog No. 50-004-18710 Supplier Medchemexpress LLC Supplier No. HYN78611ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl elaidate is a fatty acid ester derived from elaidic acid, a monounsaturated omega-9 fatty acid found in certain vegetable oils and animal fats. It is commonly used as a reference compound for the analysis of fatty acids and their derivatives by gas chromatography. This product is intended for research use only.

  • Derived from elaidic acid
  • Monounsaturated omega-9 fatty acid
  • Used as a reference compound for fatty acid analysis
  • Suitable for gas chromatography
  • Purity: 99.22%
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • For research use only
  • Store neat at 4°C under nitrogen
  • Store in solvent at -80°C for 6 months or -20°C for 1 month under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.