Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl elaidate, 95%, Thermo Scientific™

Catalog No. AAH3045803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3045803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3045803 Supplier Thermo Scientific Chemicals Supplier No. H3045803

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1937-62-8
Melting Point 9°C to 10°C
Density 0.871
Molecular Formula C19H36O2
Quantity 1 g
MDL Number MFCD00066521
Synonym methyl elaidate, elaidic acid methyl ester, methyl octadec-9-enoate, methyl trans-9-octadecenoate, e-methyl octadec-9-enoate, methyl 9-octadecenoate, 9-octadecenoic acid, methyl ester, 9-octadecenoic acid, methyl ester, e, 9-octadecenoic acid, methyl ester, 9e, methyl e-octadec-9-enoate
InChI Key QYDYPVFESGNLHU-ZHACJKMWSA-N
SMILES CCCCCCCCC=CCCCCCCCC(=O)OC
IUPAC Name methyl (E)-octadec-9-enoate
Molecular Weight (g/mol) 296.495
PubChem CID 5280590
Formula Weight 296.5
Percent Purity 95%
Chemical Name or Material Methyl elaidate
Show More Show Less
EINECSNumber 217-712-4
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.