Learn More
AdipoGen Methyl ethanesulfonate
SDP
Catalog No. 502560957
Encompass_Preferred
Chemical. CAS 1912-28-3. Formula C2H5SO3CH3. MW 124.15. Methyl ethanesulfonate is a bulding block and intermediate used in the synthesis. Methyl ethanesulfonate belongs to the group of alkyl sulfonate esters, termed as potential genotoxic impurities GTIs. Methyl ethanesulfonate may be used as reference material for the quantification of the analyte in pharmaceutical formulations using the gas chromatography technique.
Catalog No. 50-256-0957 Supplier AdipoGen Supplier No. CDXE0582M100
Chemical. CAS 1912-28-3. Formula C2H5SO3CH3. MW 124.15. Methyl ethanesulfonate is a bulding block and intermediate used in the synthesis. Methyl ethanesulfonate belongs to the group of alkyl sulfonate esters, termed as potential genotoxic impurities GTIs. Methyl ethanesulfonate may be used as reference material for the quantification of the analyte in pharmaceutical formulations using the gas chromatography technique.