Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Ethyl Ketone, ACS Reagent (2-Butanone), Reagents

Catalog No. 01337888
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-888 1 L
01-337-887 20 L
01-337-886 4 L
01-337-885 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-888 Supplier Reagents Holdings Llc Supplier No. C12505001C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl Ethyl Ketone, ACS Reagent (2-Butanone), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym 2-Butanone
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Melting Point -86.4°C
Color Colorless
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.805
Boiling Point 80°C
Grade ACS
Chemical Name or Material Methyl Ethyl Ketone, ACS Reagent (2-Butanone)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.