Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Ethyl Ketone, ASTM D740 (2-Butanone), Reagents

Catalog No. 01337174
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-174 500 mL
01-337-176 20 L
01-337-175 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-174 Supplier Reagents Holdings Llc Supplier No. C2250500500C
Add to Cart
Add to Cart

Methyl Ethyl Ketone, ASTM D740 (2-Butanone), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym 2-Butanone
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Melting Point -86.4°C
Color Colorless
Quantity 500 mL
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.805
Boiling Point 80°C
Grade Reagent
Chemical Name or Material Methyl Ethyl Ketone, ASTM D740 (2-Butanone)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.