Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Ethyl Ketone MEK ACS, Greenfield

Catalog No. 502946016
Encompass
Change view
Click to view available options
Packaging:
4x1 Fluorinated Poly Gallon Case
Metal Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
50-294-6016 Metal Pail
50-294-5973 4x1 Fluorinated Poly Gallon Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-294-6016 Supplier Thermo Scientific Production Chemicals and Services Supplier No. P354000ACSPL05M
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 78-93-3
Molecular Weight (g/mol) 72.11

Specifications

Quantity 5 Gallon
Packaging Metal Pail
Boiling Point 176 °F (80 °C)
Percent Purity 100%
Grade Reagent
Chemical Name or Material Methyl Ethyl Ketone (2-Butanone)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.