Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Ethyl Ketone, Purified, Spectrum™ Chemical
SDP

Catalog No. 18611697 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
4 L
20 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Steel Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-697 4 L Amber Glass Bottle
18-611-695 20 L Steel Can
18-611-696 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-611-697 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M12654LTGL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 78-93-3

Specifications

CAS Max % 1
Quantity 4 L
Packaging Amber Glass Bottle
Boiling Point 80°C
Grade Purified
Chemical Name or Material Methyl Ethyl Ketone

CAUTION: For manufacturing or laboratory use only. Not for Pesticide Use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.