Learn More
Medchemexpress LLC HY-N6996 100mg Medchemexpress, Methyl Eugenol CAS:93-15-2 Purity:>98%
SDP
Catalog No. 501970488
Encompass_Preferred
Medchemexpress, HY-N6996 100mg Methyl Eugenol CAS:93-15-2 Methyl Eugenol, a phenylpropanoid chemical in leaves, fruits, stems, and/or roots, may be released when that corresponding part of a plant is damaged as a result of feeding by an herbivore. Methyl Eugenol is used for male annihilation of the oriental fruit fly. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0488 Supplier Medchemexpress LLC Supplier No. HYN6996100MG
Medchemexpress, HY-N6996 100mg Methyl Eugenol CAS:93-15-2 Methyl Eugenol, a phenylpropanoid chemical in leaves, fruits, stems, and/or roots, may be released when that corresponding part of a plant is damaged as a result of feeding by an herbivore. Methyl Eugenol is used for male annihilation of the oriental fruit fly. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.