Learn More
Medchemexpress LLC Methyl Eugenol-13C,d3 | 93-15-2 | 5 MG
SDP
Catalog No. 5000419052
Encompass_Preferred
Methyl Eugenol-13C,d3 is the 13C- and deuterium-labeled form of Methyl Eugenol, a naturally occurring phenylpropanoid chemical. This compound, found in plant leaves, fruits, stems, and roots, can be released when plant parts are damaged. It is utilized in the male annihilation of the oriental fruit fly and is strictly for research purposes. The labeled compound serves as a valuable tracer and internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Stable isotope-labeled compound
  • Useful as a tracer in research
  • Functions as an internal standard for quantitative analysis
  • Derived from naturally occurring phenylpropanoid
Catalog No. 50-004-19052 Supplier Medchemexpress LLC Supplier No. HYN6996S5MG
Methyl Eugenol-13C,d3 is the 13C- and deuterium-labeled form of Methyl Eugenol, a naturally occurring phenylpropanoid chemical. This compound, found in plant leaves, fruits, stems, and roots, can be released when plant parts are damaged. It is utilized in the male annihilation of the oriental fruit fly and is strictly for research purposes. The labeled compound serves as a valuable tracer and internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Stable isotope-labeled compound
  • Useful as a tracer in research
  • Functions as an internal standard for quantitative analysis
  • Derived from naturally occurring phenylpropanoid