Learn More
Medchemexpress LLC Methyl Eugenol | 93-15-2 | 25 MG
SDP
Catalog No. 5000421708
Encompass_Preferred
Methyl Eugenol is an analytical standard primarily used in research and analytical applications. It acts as an anti-insect agent, specifically as a bait for the oriental fruit fly. This compound also exhibits anti-cancer and anti-inflammatory activities, and can induce Autophagy in cells, making it relevant for studies on intestinal ischemia/reperfusion injury.
  • Analytical standard for research and analytical applications
  • Functions as an anti-insect agent
  • Exhibits anti-cancer activities
  • Demonstrates anti-inflammatory activities
  • Induces autophagy in cells
  • Applicable in studies of intestinal ischemia/reperfusion injury
  • Suitable for HPLC, GC, and MS research experiments
Catalog No. 50-004-21708 Supplier Medchemexpress LLC Supplier No. 25MGHYN6996R
Methyl Eugenol is an analytical standard primarily used in research and analytical applications. It acts as an anti-insect agent, specifically as a bait for the oriental fruit fly. This compound also exhibits anti-cancer and anti-inflammatory activities, and can induce Autophagy in cells, making it relevant for studies on intestinal ischemia/reperfusion injury.
  • Analytical standard for research and analytical applications
  • Functions as an anti-insect agent
  • Exhibits anti-cancer activities
  • Demonstrates anti-inflammatory activities
  • Induces autophagy in cells
  • Applicable in studies of intestinal ischemia/reperfusion injury
  • Suitable for HPLC, GC, and MS research experiments