Learn More
Cayman Chemical Methyl Gallate, Purity: ≥98%, CAS: 99-24-1, 100 g
SDP
Catalog No. 502982412
Encompass_Preferred
A phenol with diverse biological activities; reduces adipogenic hormonal stimulation-induced lipid accumulation in 3T3-L1 cells at 50 µM; active against V. cholerae in vitro (MIC = 32 µg/ml) and reduces V. cholerae-induced intestinal fluid accumulation in suckling mice at 50, 100, and 200 mg/kg; reduces tumor growth and decreases the number of intratumor CD25+Foxp3high Tregs in an EL-4 model of murine lymphoma; reduces knee joint edema and leukocyte, neutrophil, and mononuclear cell infiltration in a mouse model of zymosan-induced arthritis at 0.7, 7, and 70 mg/kg
Catalog No. 50-298-2412 Supplier Cayman Chemical Supplier No. 19951100 G
A phenol with diverse biological activities; reduces adipogenic hormonal stimulation-induced lipid accumulation in 3T3-L1 cells at 50 µM; active against V. cholerae in vitro (MIC = 32 µg/ml) and reduces V. cholerae-induced intestinal fluid accumulation in suckling mice at 50, 100, and 200 mg/kg; reduces tumor growth and decreases the number of intratumor CD25+Foxp3high Tregs in an EL-4 model of murine lymphoma; reduces knee joint edema and leukocyte, neutrophil, and mononuclear cell infiltration in a mouse model of zymosan-induced arthritis at 0.7, 7, and 70 mg/kg