Learn More
Medchemexpress LLC HY-N2010 5g Medchemexpress, Methyl gallate CAS:99-24-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN20105G
Encompass_Preferred
Medchemexpress, HY-N2010 5g Methyl gallate CAS:99-24-1 Methyl gallate is a plant phenolic with antioxidant, anticancer, and anti-inflammatory activities. Methyl gallate also shows bacterial inhibition activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3870