Learn More
Selleck Chemical LLC Methyl gallate S3790-25mg
SDP
Catalog No. 503108306
Encompass_Preferred
Methyl Gallate (Methylgallate Gallic acid methyl ester Gallicin) is a plant polyphenol with antioxidant anticancer and anti-inflammatory activities
Catalog No. 50-310-8306 Supplier Selleck Chemical LLC Supplier No. S379025MG
Methyl Gallate (Methylgallate Gallic acid methyl ester Gallicin) is a plant polyphenol with antioxidant anticancer and anti-inflammatory activities