Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl germanane

Catalog No. 9091141G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-911-41G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-911-41G Supplier Sigma Aldrich Supplier No. 9091141G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Germanane is novel 2D graphene analog material. However, in sharp contrast to graphene it exhibits semiconducting properties with direct band gap of 1.50 eV. Germanane exhibits high electron mobility and its alkylation allows band gap tuning from 1.50 eV for allyl to 1.62 for methyl germanane. The semiconducting properties of alkylated germanane make them suitable for application is photocatalysis, optoelectronic, sensing etc

Specifications

CAS 88453-53-6
Molecular Formula GeCH3
Linear Formula GeCH3
MDL Number MFCD08275489
Quantity 1 g
Synonym Methyl-germylidyne; Methyl-terminated germanane
Molecular Weight (g/mol) 87.67
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.