Learn More
Accela Chembio Inc MF: C3H6O3 | MW: 90.08 | ?98% | Flash Point: 67¿C(152.6¿F) | BP: 149-151¿C | Density: 1.167g/mLat25¿C.
Catalog No. 50353624
Encompass_Preferred
MF: C3H6O3 | MW: 90.08 | ?98% | Flash Point: 67¿C(152.6¿F) | BP: 149-151¿C | Density: 1.167g/mLat25¿C.
Catalog No. 50-353-624 Supplier Accela Chembio Inc Supplier No. SY00309725G
MF: C3H6O3 | MW: 90.08 | ?98% | Flash Point: 67¿C(152.6¿F) | BP: 149-151¿C | Density: 1.167g/mLat25¿C.