Learn More
Vector Laboratories VECTOR Methyl Green Counterstain 500 ml
SDP
Catalog No. NC9374955
Encompass
Vector Methyl Green stains cell nuclei light green. This superior formulation of methyl green is suitable for use with a wide range of enzyme substrates. Optimal tissue staining is achieved by using a simple, two-step procedure. This counterstain provides an excellent alternative in multiple labeling when hematoxylin obscures the substrate colors and is suitable for use with non-aqueous mountants.
Catalog No. NC9374955 Supplier Vector Laboratories Supplier No. H3402500
Vector Methyl Green stains cell nuclei light green. This superior formulation of methyl green is suitable for use with a wide range of enzyme substrates. Optimal tissue staining is achieved by using a simple, two-step procedure. This counterstain provides an excellent alternative in multiple labeling when hematoxylin obscures the substrate colors and is suitable for use with non-aqueous mountants.