Learn More
Methyl Green-Pyronin (MGP) Solution, 250 mL, G-Biosciences
SCI_ED
Catalog No. S001111
Change view
Click to view available options
Quantity:
250 mL
Catalog No. Quantity
S001111 250 mL
1 options
Catalog No. S001111 Supplier G-Biosciences Supplier No. 7862239
Add to Cart
Add to Cart
Methyl Green-Pyronin (MGP) is used to stain plasma cells and RNA in tissue sections and cytologic preparations.
  • The solution is suitable for use with formalin fixed tissue.
  • Pyronin stains the cytoplasm of plasma cells and most nucleoli red, while Methyl green stains DNA bluish-green.
Specifications
Quantity 250 mL
Product Type Methyl Green Pyronin (Pappenheim)