Learn More
Ihc World LLC METHYL GREEN SOLUTION 250 ML
SDP
Catalog No. NC0295752
Encompass_Preferred
This Methyl Green Counterstain Solution is designed for nuclear counterstain after completion of immunohistochemical staining or for routine histology. Nuclei will be stained green color
Catalog No. NC0295752 Supplier Ihc World LLC Supplier No. IW1402
This Methyl Green Counterstain Solution is designed for nuclear counterstain after completion of immunohistochemical staining or for routine histology. Nuclei will be stained green color