Learn More
Medchemexpress LLC Methyl Green zinc chloride | 7114-03-6 | 100 MG
SDP
Catalog No. 5000410123
Encompass_Preferred
Methyl Green zinc chloride is a potent fluorescent dye. It is used as a DNA stain for cells and electrophoretic gels, and can also be used for direct measuring of viability by both microscopy and flow cytometry, with peaks at 633 and 677 nm.
  • Potent fluorescent dye.
  • Stains DNA of cells and electrophoretic gels.
  • Used for direct measuring of viability by microscopy.
  • Used for direct measuring of viability by flow cytometry.
  • Exhibits peaks at 633 and 677 nm.
Catalog No. 50-004-10123 Supplier Medchemexpress LLC Supplier No. HYD0950A500MG
Methyl Green zinc chloride is a potent fluorescent dye. It is used as a DNA stain for cells and electrophoretic gels, and can also be used for direct measuring of viability by both microscopy and flow cytometry, with peaks at 633 and 677 nm.
  • Potent fluorescent dye.
  • Stains DNA of cells and electrophoretic gels.
  • Used for direct measuring of viability by microscopy.
  • Used for direct measuring of viability by flow cytometry.
  • Exhibits peaks at 633 and 677 nm.