Learn More
Abcam Methyl green zinc chloride salt, DNA stain, 10G
Catalog No. 502704761
Encompass_Preferred
MW 653.24 g/mol. DNA stain that binds selectively to AT-rich regions of native DNA. Used as a tracking dye in acid buffers for electrophoresis. Also used in Methyl Green-Pyronin methods to demonstrate DNA and RNA in tissue sections.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4761 Supplier Abcam Supplier No. AB14628710G
MW 653.24 g/mol. DNA stain that binds selectively to AT-rich regions of native DNA. Used as a tracking dye in acid buffers for electrophoresis. Also used in Methyl Green-Pyronin methods to demonstrate DNA and RNA in tissue sections.
The product is subject to the following: Abcam Restricted Use Statement