Learn More
Medchemexpress LLC Methyl Heptadecanoate | 1731-92-6 | 284.48 | 10 MG
SDP
Catalog No. 5000447583
Encompass_Preferred
Methyl heptadecanoate is the methyl ester derivative of heptadecanoic acid. It serves as an internal standard for quantitative calibration of fatty acids in GC-MS analysis.
  • Used as an internal standard for quantitative calibration of fatty acids
  • Methyl ester derivative of heptadecanoic acid
  • White to off-white solid appearance
  • Classified as a lipid
  • Stable under recommended storage conditions
  • Soluble in DMSO
Catalog No. 50-004-47583 Supplier Medchemexpress LLC Supplier No. HYW004290S10MG
Methyl heptadecanoate is the methyl ester derivative of heptadecanoic acid. It serves as an internal standard for quantitative calibration of fatty acids in GC-MS analysis.
  • Used as an internal standard for quantitative calibration of fatty acids
  • Methyl ester derivative of heptadecanoic acid
  • White to off-white solid appearance
  • Classified as a lipid
  • Stable under recommended storage conditions
  • Soluble in DMSO