Learn More
Medchemexpress LLC Methyl heptadecanoate | 1731-92-6 | 284.48 | 50 MG
SDP
Catalog No. 5000447593
Encompass_Preferred
Methyl heptadecanoate (C17:0 methyl ester) is the methyl ester derivative of heptadecanoic acid. It is utilized as an internal standard for the quantitative calibration of fatty acids in GC-MS analysis.
  • Methyl ester derivative of heptadecanoic acid
  • Used as an internal standard for quantitative calibration
  • Suitable for GC-MS analysis
Catalog No. 50-004-47593 Supplier Medchemexpress LLC Supplier No. HYW004290S50MG
Methyl heptadecanoate (C17:0 methyl ester) is the methyl ester derivative of heptadecanoic acid. It is utilized as an internal standard for the quantitative calibration of fatty acids in GC-MS analysis.
  • Methyl ester derivative of heptadecanoic acid
  • Used as an internal standard for quantitative calibration
  • Suitable for GC-MS analysis