Learn More
Medchemexpress LLC Methyl heptadecanoate | 1731-92-6 | 284.48 | 500 MG
SDP
Catalog No. 5000448574
Encompass_Preferred
Methyl heptadecanoate (C17:0 methyl ester) is the methyl ester derivative of heptadecanoic acid. It is utilized as an internal standard for the quantitative calibration of fatty acids in GC-MS analysis.
  • Used as an internal standard for quantitative calibration of fatty acids in GC-MS analysis.
Catalog No. 50-004-48574 Supplier Medchemexpress LLC Supplier No. HYW004290500MG
Methyl heptadecanoate (C17:0 methyl ester) is the methyl ester derivative of heptadecanoic acid. It is utilized as an internal standard for the quantitative calibration of fatty acids in GC-MS analysis.
  • Used as an internal standard for quantitative calibration of fatty acids in GC-MS analysis.