Learn More
Medchemexpress LLC Methyl heptadecanoate-d33 | 1219804-81-5 | 317.68 | 25 MG
SDP
Catalog No. 5000447743
Encompass_Preferred
Methyl heptadecanoate-d33 is a deuterium-labeled form of methyl heptadecanoate, an ester derivative of heptadecanoic acid. It serves as an internal standard for the quantitative calibration of fatty acids in GC-MS analysis. Deuterium labeling is also notable for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Deuterium labeling can impact pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-47743 Supplier Medchemexpress LLC Supplier No. 25MGHYW004290S
Add to Cart
Add to Cart
Methyl heptadecanoate-d33 is a deuterium-labeled form of methyl heptadecanoate, an ester derivative of heptadecanoic acid. It serves as an internal standard for the quantitative calibration of fatty acids in GC-MS analysis. Deuterium labeling is also notable for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Deuterium labeling can impact pharmacokinetic and metabolic profiles of drugs